missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 169835000

 View more versions of this product

Catalog No. AC169835000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diiodomethane
6.0°C
3.3200g/mL
>110°C
99% min. (GC)
CH2I2
CH2I2
500 g
09,170; 11,182; 17,116
15,6137
Solubility in water: insoluble. Other solubilities: miscible with alcohol,propanol,isopropanol,,hexane,cyclohexane,ether,chloroform and benzene
ICI
267.84
6346
267.83
Liquid
75-11-6,7440-22-4
Red to Yellow
181.0°C
Authentic
Glass bottle
1.737
MFCD00001079
01,71
3.32
methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane
NZZFYRREKKOMAT-UHFFFAOYSA-N
diiodomethane
Silver wire
2.80 mPa.s (20°C)
99+%

Chemical Identifiers

75-11-6
267.84
NZZFYRREKKOMAT-UHFFFAOYSA-N
6346
ICI
CH2I2
MFCD00001079
methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane
diiodomethane
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Danger

EINECSNumber : 200-841-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only