missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diiodomethane, 99%, stab., Alfa Aesar™

Manufacturer:  Alfa Aesar A1545736

 View more versions of this product

Catalog No. AAA1545736



Description

Description

Applications
Diiodomethane is used to determine the density of minerals and other solid samples due to its high density. It is used as an optical contact liquid to determine the refractive index of certain gemstones. In simmons-smith reaction, it acts as a reagent to generate methylene (carbine) free radical. It reacts with olefins to prepare cyclopropanes with high stereo specificity.

Solubility
Miscible with alcohol and ether. Slightly miscible with water.

Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with alkali metal salts, strong oxidizing agents, strong bases, metals, lithium, potassium, sodium, sodium oxides and alloys.
Specifications

Specifications

Diiodomethane
5°C to 6°C
stabilized
99%, stab.
MFCD00001079
1696892
methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane
C(I)I
267.836
267.84
99%
180°C to 181°C (decomposition)
500g
75-11-6
CH2I2
UN2810
14,6066
NZZFYRREKKOMAT-UHFFFAOYSA-N
diiodomethane
6346
Liquid

Chemical Identifiers

75-11-6
267.836
NZZFYRREKKOMAT-UHFFFAOYSA-N
6346
C(I)I
CH2I2
MFCD00001079
methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane
diiodomethane
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

RTECSNumber : PA8575000

TSCA : TSCA

Documents