Please login to your online account to display your discounted pricing

Alfa Aesar™ Diiodofluorescein

$51.88 - $163.16

Chemical Identifiers

Chemical Name or Material Diiodofluorescein
CAS 31395-16-1
Molecular Formula C20H10I2O5
Formula Weight 584.11
MDL Number MFCD00036461
Synonym C.I. 45425
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1655409

 
alfa aesar™
A1655409
10g Each for $51.88

AAA1655418

 
alfa aesar™
A1655418
50g Each for $163.16
Description & Specifications

Specifications

Formula Weight 584.11
Chemical Name or Material Diiodofluorescein
Melting Point 239° to 241°C (dec.)
Molecular Formula C20H10I2O5
Quantity 10g
MDL Number MFCD00036461
CAS 31395-16-1
Synonym C.I. 45425