missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diiodobis(triphenylphosphine)nickel(II)

Manufacturer:  Alfa Aesar™ 1886904

 View more versions of this product

Catalog No. AA1886904



Specifications

Specifications

Diiodobis(triphenylphosphine)nickel(II)
Brown
82308-17-6
[(C6H5)3P]2NiI2
diiododbis triphenylphosphinenickel ii, diiodobis triphenylphosphine nickel ii, nii2 pph3 2, diiodonickel; triphenylphosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Ni](I)I
837.086
837.08
Air & Light Sensitive
2g
C36H30I2NiP2
MFCD00015866
AKXNPPNNEDIAJL-UHFFFAOYSA-L
diiodonickel;triphenylphosphane
6102125
Crystalline Powder

Chemical Identifiers

82308-17-6
837.086
AKXNPPNNEDIAJL-UHFFFAOYSA-L
6102125
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Ni](I)I
C36H30I2NiP2
MFCD00015866
diiododbis triphenylphosphinenickel ii, diiodobis triphenylphosphine nickel ii, nii2 pph3 2, diiodonickel; triphenylphosphane
diiodonickel;triphenylphosphane
Safety and Handling

Safety and Handling

GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

SDS Documents