Please login to your online account to display your discounted pricing

Alfa Aesar™ Diiodobis(triphenylphosphine)nickel(II)

$60.66 - $195.49

Chemical Identifiers

Chemical Name or Material Diiodobis(triphenylphosphine)nickel(II)
CAS 82308-17-6
Linear Formula [(C6H5)3P]2NiI2
Formula Weight 837.08
MDL Number MFCD00015866
Synonym Bis(triphenylphosphine)nickel(II) iodide; Bis(triphenylphosphine)diiodonickel(II)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1886904

 
alfa aesar™
1886904
2g Each for $60.66

AA1886909

 
alfa aesar™
1886909
10g Each for $195.49
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 837.08
Chemical Name or Material Diiodobis(triphenylphosphine)nickel(II)
Color Dark brown
Quantity 2g
Linear Formula [(C6H5)3P]2NiI2
MDL Number MFCD00015866
CAS 82308-17-6
Sensitivity Air & Light Sensitive
Synonym Bis(triphenylphosphine)nickel(II) iodide; Bis(triphenylphosphine)diiodonickel(II)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.