Learn More
Diiodo(p-cymene)ruthenium(II) dimer, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04688606
Description
Diiodo(p-cymene)ruthenium(II) dimer can be applied as a catalyst for many reactions such as ruthenium-catalyzed hydrogenation, preparation of Walphos-type ferrocenyl-aryl diphosphine ligands, ruthenium-catalyzed synthesis reactions, and enantioselective hydrogenation of ketones and olefins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diiodo(p-cymene)ruthenium(II) dimer | |
Brown-Yellow | |
5 g | |
Di-μ-iodobis[(p-cymene)iodoruthenium(II)] | |
978.2 |
90614-07-6 | |
MFCD08064027 | |
Light and moisture sensitive | |
Insoluble in water. | |
Crystalline Solid |
Chemical Identifiers
90614-07-6 | |
MFCD08064027 | |
Di-μ-iodobis[(p-cymene)iodoruthenium(II)] |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P264b-P270-P280i-P281-P301+P312-P305+P351+P338-P308+P313-P330-P501c
H302-H319-H341-H500
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only