Please login to your online account to display your discounted pricing

Alfa Aesar™ Diiodo[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II)

Chemical Identifiers

Chemical Name or Material Diiodo[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II)
Formula Weight 834.66
MDL Number MFCD14155730
Synonym PdI2(dtbpf); [1,1'-Bis(di-tert-butylphosphino)ferrocene]diiodopalladium(II)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4690203

  SDS
alfa aesar™
4690203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 834.66
Physical Form Crystalline Solid
Sensitivity Air & Light Sensitive
MDL Number MFCD14155730
Synonym PdI2(dtbpf); [1,1'-Bis(di-tert-butylphosphino)ferrocene]diiodopalladium(II)
Color Dark brown
Quantity 1g
Chemical Name or Material Diiodo[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II)