Learn More
Dihydrogen hexahydroxyplatinate(IV), 99.9% (metals basis), Pt 61.0% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01105202
Description
Dihydrogen hexahydroxyplatinate(IV) is used as a precursor to produce platinum dioxide (II), a catalyst of hydrogenation and hydrogenolysis of carbonyl group or C-Cl linkages in acyl chlorides. It also acts as a precursor to produce other platinum compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dihydrogen hexahydroxyplatinate(IV) | |
(decomposition) | |
(metals basis) | |
MFCD00058744 | |
Hygroscopic | |
Insoluble in water. | |
[H+].[H+].[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Pt+4] | |
299.14 | |
99.9% | |
Crystalline |
51850-20-5 | |
Yellow | |
H8O6Pt | |
0.5 g | |
Hexahydroxyplatinic acid; Hydrogen hexahydroxyplatinate(IV) | |
ILXDKMTYQJUXAB-UHFFFAOYSA-J | |
platinum(4+) dihydrogen hexahydroxide | |
299.17 | |
Pt ≤61.0% |
Chemical Identifiers
51850-20-5 | |
299.14 | |
ILXDKMTYQJUXAB-UHFFFAOYSA-J | |
platinum(4+) dihydrogen hexahydroxide |
H8O6Pt | |
MFCD00058744 | |
Hexahydroxyplatinic acid; Hydrogen hexahydroxyplatinate(IV) | |
[H+].[H+].[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Pt+4] |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
H315-H317-H319-H335
EINECSNumber : 257-471-2
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only