Please login to your online account to display your discounted pricing

Alfa Aesar™ Dihydrogen hexachloroiridate(IV) hydrate, 99% (metals basis), Ir 38-42%

Chemical Identifiers

Chemical Name or Material Dihydrogen hexachloroiridate(IV) hydrate
CAS 110802-84-1
Formula Weight 406.93 (Anhydrous)
MDL Number MFCD00011328
UN Number UN3260
Synonym Hydrogen hexachloroiridate(IV); Chloroiridic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1103103

  SDS
alfa aesar™
1103103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1103106

  SDS
alfa aesar™
1103106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1103114

  SDS
alfa aesar™
1103114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 406.93 (Anhydrous)
Physical Form Lumps
TSCA TSCA
UN Number UN3260
Assay 99%
Health Hazard 2 GHS H Statement
H314-H318-H290-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
Concentration Ir 38 to 42%
MDL Number MFCD00011328
Solubility Information Soluble in alcohol and water
Chemical Name or Material Dihydrogen hexachloroiridate(IV) hydrate
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Quantity 1g
Color Black
CAS 110802-84-1
Synonym Hydrogen hexachloroiridate(IV); Chloroiridic acid
Assay Percent Notes (metals basis)
RTECS Number NO3592500
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H290-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.