missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dihydrogen hexachloroiridate(IV) hydrate, 99% (metals basis), Ir 38-42%

Manufacturer:  Alfa Aesar™ 1103114

 View more versions of this product

Catalog No. AA1103114



Specifications

Specifications

Dihydrogen hexachloroiridate(IV) hydrate
110802-84-1
Cl6H4IrO
UN3260
Ir 38 to 42%
YDVQBPXDKJKDME-UHFFFAOYSA-J
tetrachloroiridium;hydrate;dihydrochloride
16211483
Lumps
Ir 38 to 42%
25g
(metals basis)
MFCD00011328
Hydrogen hexachloroiridate(IV); Chloroiridic acid
Soluble in alcohol and water
O.Cl.Cl.Cl[Ir](Cl)(Cl)Cl
424.948
406.93 (Anhydrous)
99%
Black

Chemical Identifiers

110802-84-1
424.948
YDVQBPXDKJKDME-UHFFFAOYSA-J
16211483
O.Cl.Cl.Cl[Ir](Cl)(Cl)Cl
Cl6H4IrO
MFCD00011328
Hydrogen hexachloroiridate(IV); Chloroiridic acid
tetrachloroiridium;hydrate;dihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H290-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

RTECSNumber : NO3592500

TSCA : TSCA

Documents