Please login to your online account to display your discounted pricing

Alfa Aesar™ Dihydrogen hexabromoplatinate(IV) nonahydrate, 99.9% (metals basis), Pt 23.7% min

Chemical Identifiers

Chemical Name or Material Dihydrogen hexabromoplatinate(IV) nonahydrate
CAS 20596-34-3
Linear Formula H2PtBr6·9H2O
Formula Weight 838.67 (676.56 Anhydrous)
MDL Number MFCD00049645
UN Number UN3260
Synonym Bromoplatinic acid; Platinic bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1261703

  SDS
alfa aesar™
1261703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1261706

  SDS
alfa aesar™
1261706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Dihydrogen hexabromoplatinate(IV) nonahydrate
Concentration Pt 23.7% min.
Quantity 1g
CAS 20596-34-3
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P260-P284a-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
TSCA TSCA
Physical Form Crystalline
Assay 99.9%
Assay Percent Notes (metals basis)
Linear Formula H2PtBr6·9H2O
Color Red-brown
Formula Weight 838.67 (676.56 Anhydrous)
MDL Number MFCD00049645
UN Number UN3260
Synonym Bromoplatinic acid; Platinic bromide
Safety and Handling

Danger

GHS P Statement
P260-P284a-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.