missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dihydrogen hexabromoplatinate(IV) nonahydrate, 99.9% (metals basis), Pt 23.7% min

Manufacturer:  Alfa Aesar™ 1261706

 View more versions of this product

Catalog No. AA1261706



Specifications

Specifications

Dihydrogen hexabromoplatinate(IV) nonahydrate
5g
(metals basis)
MFCD00049645
Bromoplatinic acid; Platinic bromide
838.67 (676.56 Anhydrous)
99.9%
Red to Brown
20596-34-3
H2PtBr6·9H2O
UN3260
Pt 23.7% min.
Crystalline

Chemical Identifiers

20596-34-3
MFCD00049645
Bromoplatinic acid; Platinic bromide
Safety and Handling

Safety and Handling

GHS H Statement
H334-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P284a-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Documents