Please login to your online account to display your discounted pricing

Dihydrogen dinitrosulfatoplatinate(II) solution, Pt 4-6% (cont. Pt), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Dihydrogen dinitrosulfatoplatinate(II) solution
Name Note Pt 4-6% (cont. Pt)
CAS 12033-81-7
Physical Form Liquid
Molecular Formula H2Pt(NO2)2SO4
Formula Weight 384.17 (Anhydrous)
MDL Number MFCD00064668
Synonym Dinitrosulfatoplatinum(II)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA12627NA

  SDS
alfa aesar
12627NA
≈2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA12627NC

  SDS
alfa aesar
12627NC
≈10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA12627NJ

  SDS
alfa aesar
12627NJ
≈0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 384.17 (anhydrous)
Quantity ≈2g
Color Brown
DOT Information DOT Class: 8, Packaging Group: III
Chemical Name or Material Dihydrogen dinitrosulfatoplatinate(II) solution
Name Note Pt 4-6% (cont. Pt)
Physical Form Liquid
Molecular Formula H2Pt(NO2)2SO4
Appearance Brown liquid
MDL Number MFCD00064668
CAS 12033-81-7
Synonym Dinitrosulfatoplatinum(II)
Health Hazard 1 H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.
Safety and Handling

H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.