Please login to your online account to display your discounted pricing

Alfa Aesar™ Dihydroergocristine methanesulfonate

5-HT receptor antagonist; partial agonist at adrenergic and dopaminergic receptors

Chemical Identifiers

Chemical Name or Material Dihydroergocristine methanesulfonate
CAS 24730-10-7
Molecular Formula C36H45N5O8S
Formula Weight 707.8
MDL Number MFCD00153792
Merck Index 14,3653
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ60638EX3

 
alfa aesar™
J60638EX3
200mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 707.8
Molecular Formula C36H45N5O8S
CAS 24730-10-7
Chemical Name or Material Dihydroergocristine methanesulfonate
Physical Form Powder
Color White
MDL Number MFCD00153792
Merck Index 14,3653
RTECS Number KE7600000
Quantity 200mg