Please login to your online account to display your discounted pricing

Alfa Aesar™ Dihydroceramide C8 , 98%


Quantity 5mg

Manufacturer: alfa aesar™  J67477LB0

 View more versions of this product

Catalog No. AAJ67477LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 427.71
CAS 145774-33-0
MDL Number MFCD01320385
Physical Form Waxy Solid
Assay 98%
Chemical Name or Material Dihydroceramide C8
Storage Note 1 -30° to -10°C
Synonym D-erythro-N-Octanoyldihydrosphingosine
Color White
Quantity 5mg

Specifications

Chemical Name or Material Dihydroceramide C8
CAS 145774-33-0
Formula Weight 427.71
MDL Number MFCD01320385
Synonym D-erythro-N-Octanoyldihydrosphingosine
Accessories

-30° to -10°C