Please login to your online account to display your discounted pricing

Alfa Aesar™ Dihydroceramide C8 , 98%

Chemical Identifiers

Chemical Name or Material Dihydroceramide C8
CAS 145774-33-0
Formula Weight 427.71
MDL Number MFCD01320385
Synonym D-erythro-N-Octanoyldihydrosphingosine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67477LB0

 
alfa aesar™
J67477LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 427.71
Chemical Name or Material Dihydroceramide C8
Physical Form Waxy Solid
Assay 98%
MDL Number MFCD01320385
CAS 145774-33-0
Color White
Synonym D-erythro-N-Octanoyldihydrosphingosine
Storage Note 1 -30° to -10°C
Quantity 5mg
Safety and Handling

-30° to -10°C