Please login to your online account to display your discounted pricing

Dihydridotetrakis(triphenylphosphine)ruthenium(II), 97%, ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC367495000

 View more versions of this product

Catalog No. AC367495000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0% (ex Ru)
Color Ochre
Physical Form Lustrous Powder
Formula Weight 1152.25
Quantity 500mg
Synonym Tetrakis(triphenylphosphine)ruthenium(II) hydride
Linear Formula [(C6H5)3P]4Ru
MDL Number MFCD04126429
CAS 19529-00-1
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C72H62P4Ru
Chemical Name or Material Dihydridotetrakis(triphenylphosphine)ruthenium(II)
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Dihydridotetrakis(triphenylphosphine)ruthenium(II)
Assay ≥96.0% (ex Ru)
Assay Percent Range 97%
Molecular Formula C72H62P4Ru
Linear Formula [(C6H5)3P]4Ru
Formula Weight 1152.25
MDL Number MFCD04126429
Synonym Tetrakis(triphenylphosphine)ruthenium(II) hydride
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents