Please login to your online account to display your discounted pricing

Dihydridotetrakis(triphenylphosphine)ruthenium(II), 97%, ACROS Organics™

$185.30 - $185.30

Chemical Identifiers

Chemical Name or Material Dihydridotetrakis(triphenylphosphine)ruthenium(II)
Assay ≥96.0% (ex Ru)
Assay Percent Range 97%
Molecular Formula C72H62P4Ru
Linear Formula [(C6H5)3P]4Ru
Formula Weight 1152.25
MDL Number MFCD04126429
Synonym Tetrakis(triphenylphosphine)ruthenium(II) hydride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC367495000

  SDS
thermo fisher scientific
AC367495000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC367490050

  SDS
thermo fisher scientific
AC367490050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC367490010

  SDS
thermo fisher scientific
AC367490010
Glass bottle 1g Each for $185.30
Description & Specifications

Specifications

Assay ≥96.0% (ex Ru)
Physical Form Lustrous Powder
Color Ochre
Formula Weight 1152.25
Quantity 500mg
Synonym Tetrakis(triphenylphosphine)ruthenium(II) hydride
Packaging Glass bottle
Linear Formula [(C6H5)3P]4Ru
MDL Number MFCD04126429
CAS 19529-00-1
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Chemical Name or Material Dihydridotetrakis(triphenylphosphine)ruthenium(II)
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C72H62P4Ru
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Reduction reagent

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.