missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diglycolic anhydride, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 114671000
Specifications
Diglycolic anhydride | |
Glass bottle | |
Cream to White | |
100g | |
4480-83-5 | |
C4H4O4 | |
19, 153 | |
Solubility in water: soluble | |
C1C(=O)OC(=O)CO1 | |
116.07 | |
116.07 | |
97% |
Authentic | |
240°C to 241°C | |
90°C to 95°C | |
97% | |
97% | |
MFCD00006677 | |
4-maleimidobutyric acid hydrazide hydrochloride, acetic acid, oxydi-, cyclic anhydride, diglycolic anhydride, diglycolicanhydride, digylcolic anhydride, digylcolicanhydride, oxydiacetic anhydride, p-dioxane-2,6-dione, pubchem24315, r-1-methyl-2-pyrrolidineethanamine dihydrochloride | |
PIYNUZCGMLCXKJ-UHFFFAOYSA-N | |
1,4-dioxane-2,6-dione | |
78232 | |
Crystalline Powder, Crystals or Needles |
Chemical Identifiers
4480-83-5 | |
116.07 | |
PIYNUZCGMLCXKJ-UHFFFAOYSA-N | |
78232 | |
C1C(=O)OC(=O)CO1 |
C4H4O4 | |
MFCD00006677 | |
4-maleimidobutyric acid hydrazide hydrochloride, acetic acid, oxydi-, cyclic anhydride, diglycolic anhydride, diglycolicanhydride, digylcolic anhydride, digylcolicanhydride, oxydiacetic anhydride, p-dioxane-2,6-dione, pubchem24315, r-1-methyl-2-pyrrolidineethanamine dihydrochloride | |
1,4-dioxane-2,6-dione |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
EINECSNumber : 224-761-5