missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diglycolic anhydride, 97%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 114670250

 View more versions of this product

Catalog No. AC114670250



Description

Description

Specifications

Specifications

Diglycolic anhydride
Glass bottle
Cream to White
25g
4480-83-5
C4H4O4
19, 153
Solubility in water: soluble
C1C(=O)OC(=O)CO1
116.07
116.07
97%
Authentic
240°C to 241°C
90°C to 95°C
97%
97%
MFCD00006677
4-maleimidobutyric acid hydrazide hydrochloride, acetic acid, oxydi-, cyclic anhydride, diglycolic anhydride, diglycolicanhydride, digylcolic anhydride, digylcolicanhydride, oxydiacetic anhydride, p-dioxane-2,6-dione, pubchem24315, r-1-methyl-2-pyrrolidineethanamine dihydrochloride
PIYNUZCGMLCXKJ-UHFFFAOYSA-N
1,4-dioxane-2,6-dione
78232
Crystalline Powder, Crystals or Needles

Chemical Identifiers

4480-83-5
116.07
PIYNUZCGMLCXKJ-UHFFFAOYSA-N
78232
C1C(=O)OC(=O)CO1
C4H4O4
MFCD00006677
4-maleimidobutyric acid hydrazide hydrochloride, acetic acid, oxydi-, cyclic anhydride, diglycolic anhydride, diglycolicanhydride, digylcolic anhydride, digylcolicanhydride, oxydiacetic anhydride, p-dioxane-2,6-dione, pubchem24315, r-1-methyl-2-pyrrolidineethanamine dihydrochloride
1,4-dioxane-2,6-dione
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 224-761-5

SDS Documents
Product Certifications