Digital Timer with Photogates
Microprocessor-controlled four-digit timer.
$454.15 - $454.15
Specifications
For Use With (Application) Air tracks, free-fall apparatus and other equipment
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Equipment) Includes Product Type Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Includes Product Type Price Quantity  
S05928 Fisher Scientific
DTPHG1
Air tracks, free-fall apparatus and other equipment Unit Indicators, function indicator, on/off switch, free fall switch, stop button, data display, function selector, clear/zero button, 2 photogates with support rods and screws Digital Timer Each for $454.15
Specifications
Description
Designed to be used with air tracks, free-fall apparatus and other equipment.
  • Offers eight programmed functions to record counts, time intervals, periods of oscillation, accelerations, acceleration due to gravity and velocities in collisions
  • Comes with two photogates
Description & Specifications
Specifications
Air tracks, free-fall apparatus and other equipment