Binder™ Digital Pressure Display
Binder™ Digital Pressure Display had a measuring range from 1mbar to atmospheric pressure.
Manufacturer:  Binder™ 80120806
 View more versions of this product
Catalog No. 08111198
Description
Description
  • Display accuracy of 1mbar
  • For use with either VD 53 or VD 115
Specifications
Specifications
VD 53
From 1mbar to atmospheric pressure
Digital pressure display
Documents