Please login to your online account to display your discounted pricing

Digital Meters


Designed for simple, accurate test readings.

Manufacturer: fisher science education  1845

 View more versions of this product

Catalog No. S02337

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Specifications

Includes Printed instructions and plant pH preference list packed with the meter
For Use With (Application) pH
Detection Range 3.5 to 9pH