missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Difurfurylsulfide, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 265080050

 View more versions of this product

Catalog No. AC265080050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Difurfurylsulfide
Authentic
1.5540 to 1.5560
White to Brown
5g
95% min. (GC)
17,III,355
Solubility in water: insoluble
C1=COC(=C1)CSCC2=CC=CO2
194.25
194.25
96%
>110°C
Glass bottle
135.0°C to 143.0°C (14.0mmHg)
30.0°C to 32.0°C
13678-67-6
C10H10SO2
2-difurfuryl sulfide, 2-furfuryl monosulfide, bis 2-furfuryl sulfide, bis furan-2-ylmethyl sulfane, bis-furfuryl sulfide, difurfuryl sulfide, difurfurylsulfide, furan, 2,2'-thiobis methylene bis, furfuryl sulfide, unii-x6o686cpco
UYLKDZXJEKFFHJ-UHFFFAOYSA-N
2-(furan-2-ylmethylsulfanylmethyl)furan
61659
Low Melting Solid or Liquid

Chemical Identifiers

13678-67-6
194.25
2-difurfuryl sulfide, 2-furfuryl monosulfide, bis 2-furfuryl sulfide, bis furan-2-ylmethyl sulfane, bis-furfuryl sulfide, difurfuryl sulfide, difurfurylsulfide, furan, 2,2'-thiobis methylene bis, furfuryl sulfide, unii-x6o686cpco
2-(furan-2-ylmethylsulfanylmethyl)furan
C10H10SO2
UYLKDZXJEKFFHJ-UHFFFAOYSA-N
61659
C1=COC(=C1)CSCC2=CC=CO2
Safety and Handling

Safety and Handling

EINECSNumber : 237-172-3

SDS Documents