missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Difurfurylsulfide, 96%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC265080250
Specifications
Difurfurylsulfide | |
Authentic | |
1.5540 to 1.5560 | |
White to Brown | |
25g | |
95% min. (GC) | |
17,III,355 | |
Solubility in water: insoluble | |
C1=COC(=C1)CSCC2=CC=CO2 | |
194.25 | |
194.25 | |
96% |
>110°C | |
Glass bottle | |
135.0°C to 143.0°C (14.0mmHg) | |
30.0°C to 32.0°C | |
13678-67-6 | |
C10H10SO2 | |
2-difurfuryl sulfide, 2-furfuryl monosulfide, bis 2-furfuryl sulfide, bis furan-2-ylmethyl sulfane, bis-furfuryl sulfide, difurfuryl sulfide, difurfurylsulfide, furan, 2,2'-thiobis methylene bis, furfuryl sulfide, unii-x6o686cpco | |
UYLKDZXJEKFFHJ-UHFFFAOYSA-N | |
2-(furan-2-ylmethylsulfanylmethyl)furan | |
61659 | |
Low Melting Solid or Liquid |
Chemical Identifiers
13678-67-6 | |
194.25 | |
2-difurfuryl sulfide, 2-furfuryl monosulfide, bis 2-furfuryl sulfide, bis furan-2-ylmethyl sulfane, bis-furfuryl sulfide, difurfuryl sulfide, difurfurylsulfide, furan, 2,2'-thiobis methylene bis, furfuryl sulfide, unii-x6o686cpco | |
2-(furan-2-ylmethylsulfanylmethyl)furan |
C10H10SO2 | |
UYLKDZXJEKFFHJ-UHFFFAOYSA-N | |
61659 | |
C1=COC(=C1)CSCC2=CC=CO2 |
Safety and Handling
EINECSNumber : 237-172-3