Please login to your online account to display your discounted pricing

Alfa Aesar™ Difluoroacetic acid, 98%

$75.28 - $75.28

Chemical Identifiers

Chemical Name or Material Difluoroacetic acid
CAS 381-73-7
Molecular Formula F2CHCO2H
Formula Weight 96.03
MDL Number MFCD00004220
UN Number UN3265
Beilstein 1098588
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1278706

 
alfa aesar™
A1278706
5g Each for $75.28
Description & Specifications

Specifications

Formula Weight 96.03
Chemical Name or Material Difluoroacetic acid
TSCA TSCA
Assay 98%
Molecular Formula F2CHCO2H
MDL Number MFCD00004220
CAS 381-73-7
UN Number UN3265
Beilstein 1098588
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
RTECS Number AG9900000
Flash Point 78°C (172°F)
Odor Pungent
Boiling Point 132° to 134°C
Melting Point −1°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.