missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethylzinc, nominally 15% w/w in hexanes, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 4488014

 View more versions of this product

Catalog No. AA4488014



Description

Description

  • Packaged under Argon
Specifications

Specifications

Diethylzinc
557-20-0
−40°C (−40°F)
Brown to Colorless
Air & Moisture Sensitive
Zn(C2H5)2
UN3394
IPSRAFUHLHIWAR-UHFFFAOYSA-N
zinc;ethane
11185
Liquid
Nominally 15% w/w in hexanes
25g
0.720g/mL
ChemSeal™ bottles
C4H10Zn
MFCD00009021
diethylzinc, zinc, diethyl, diethyl zinc, zinc ethide, zinc ethyl, unii-s0w5nqh7c6, et2zn, znet2, s0w5nqh7c6, c2h5 2zn
C[CH2-].C[CH2-].[Zn+2]
123.504
123.49

Chemical Identifiers

557-20-0
123.504
IPSRAFUHLHIWAR-UHFFFAOYSA-N
11185
C[CH2-].C[CH2-].[Zn+2]
C4H10Zn
MFCD00009021
diethylzinc, zinc, diethyl, diethyl zinc, zinc ethide, zinc ethyl, unii-s0w5nqh7c6, et2zn, znet2, s0w5nqh7c6, c2h5 2zn
zinc;ethane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H250-H260-H252-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities
may catch fire.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

SDS Documents