Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethylzinc, nominally 15% w/w in hexanes, packaged under Argon in resealable ChemSeal™ bottles

$68.86 - $68.86

Chemical Identifiers

Chemical Name or Material Diethylzinc
CAS 557-20-0
Molecular Formula C4H10Zn
Linear Formula Zn(C2H5)2
Formula Weight 123.49
MDL Number MFCD00009021
UN Number UN3394
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4488014

  SDS
alfa aesar™
4488014
25g Each for $68.86

AA4488022

  SDS
alfa aesar™
4488022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Diethylzinc
Concentration Nominally 15% w/w in hexanes
CAS 557-20-0
Health Hazard 1 Danger
Quantity 25g
Flash Point −40°C (−40°F)
Health Hazard 2 GHS H Statement
H225-H250-H260-H252-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities
may catch fire.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 0.720g/mL
Storage Note 1
  • Packaged under Argon
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
Color Colorless to pale brown
Packaging ChemSeal™ bottles
Sensitivity Air & Moisture Sensitive
TSCA TSCA
Physical Form Liquid
Molecular Formula C4H10Zn
Linear Formula Zn(C2H5)2
Formula Weight 123.49
MDL Number MFCD00009021
UN Number UN3394

  • Packaged under Argon

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H250-H260-H252-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities
may catch fire.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.

  • Packaged under Argon