Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethylzinc, 96%

$387.89 - $387.89

Chemical Identifiers

Chemical Name or Material Diethylzinc
CAS 557-20-0
Linear Formula Zn(C2H5)2
Formula Weight 123.49
MDL Number MFCD00009021
UN Number UN3394
Merck Index 14,3131
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3956422

 
alfa aesar™
3956422
100g Each for $387.89
Description & Specifications

Specifications

Formula Weight 123.49
Health Hazard 3 GHS P Statement
P210-P273-P305+P351+P338-P309-P310-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Diethylzinc
Sensitivity Air & Moisture Sensitive
TSCA TSCA
Melting Point −28°C
Physical Form Liquid
Assay 96%
Linear Formula Zn(C2H5)2
MDL Number MFCD00009021
CAS 557-20-0
UN Number UN3394
Merck Index 14,3131
Health Hazard 1 Danger
Quantity 100g
Flash Point −18° (−1°F)
Health Hazard 2 GHS H Statement
H250-H260-H314-H318
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Density 1.18g/mL
Boiling Point 124°C
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
Color Colorless
Packaging Cylinder
Safety and Handling

Danger

GHS P Statement
P210-P273-P305+P351+P338-P309-P310-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H250-H260-H314-H318
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.