missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyltin dichloride

Manufacturer:  Alfa Aesar™ 7115504

 View more versions of this product

Catalog No. AA7115504



Specifications

Specifications

Diethyltin dichloride
84°C
2g
Moisture Sensitive
(C2H5)2SnCl2
diethyltin dichloride, diethyldichlorotin, diethyltindichloride, dichlorodiethyltin, diethyldichlorostannane, tin, dichlorodiethyl, stannane, dichlorodiethyl, diethyltin chloride, tin, diethyl-, dichloride, unii-pga900d045
CC[Sn](CC)(Cl)Cl
247.734
247.72
220°C
866-55-7
White
C4H10Cl2Sn
MFCD00013609
YFAXVVMIXZAKSR-UHFFFAOYSA-L
dichloro(diethyl)stannane
91491
Crystalline

Chemical Identifiers

866-55-7
247.734
YFAXVVMIXZAKSR-UHFFFAOYSA-L
91491
CC[Sn](CC)(Cl)Cl
C4H10Cl2Sn
MFCD00013609
diethyltin dichloride, diethyldichlorotin, diethyltindichloride, dichlorodiethyltin, diethyldichlorostannane, tin, dichlorodiethyl, stannane, dichlorodiethyl, diethyltin chloride, tin, diethyl-, dichloride, unii-pga900d045
dichloro(diethyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : WH7200000

SDS Documents