Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethylphenylphosphine, 97%

Chemical Identifiers

Chemical Name or Material Diethylphenylphosphine
CAS 1605-53-4
Formula Weight 166.21
MDL Number MFCD00015172
UN Number UN3278
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1018003

  SDS
alfa aesar™
1018003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1018006

  SDS
alfa aesar™
1018006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1018014

  SDS
alfa aesar™
1018014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 166.21
Chemical Name or Material Diethylphenylphosphine
Physical Form Liquid
Assay 97%
MDL Number MFCD00015172
CAS 1605-53-4
UN Number UN3278
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H227-H302-H312
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P210-P261-P280-P304+P340-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Refractive Index 1.5475
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
Density 0.954g/mL
Flash Point 66° (150°F)
Odor Pungent
Sensitivity Air & Moisture Sensitive
Boiling Point 220°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P304+P340-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H227-H302-H312
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.