Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethylmethylvinylsilane, 98+%

Chemical Identifiers

Chemical Name or Material Diethylmethylvinylsilane
CAS 18292-29-0
Molecular Formula C7H16Si
Linear Formula (CH3CH2)2CH3SiCH=CH2
Formula Weight 128.29
MDL Number MFCD00041344
UN Number UN1993
Synonym Vinyldiethylmethylsilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2321406

  SDS
alfa aesar™
B2321406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 128.29
Quantity 5g
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapour.
Assay ≥98%
Molecular Formula C7H16Si
Chemical Name or Material Diethylmethylvinylsilane
Boiling Point 118° to 119°C
MDL Number MFCD00041344
Linear Formula (CH3CH2)2CH3SiCH=CH2
Health Hazard 1 Danger
CAS 18292-29-0
UN Number UN1993
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym Vinyldiethylmethylsilane
Flash Point 1°C (35°F)
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapour.