Please login to your online account to display your discounted pricing

Diethylmethoxyborane, 4M solution in THF, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC398388000

 View more versions of this product

Catalog No. AC398388000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. Handle under inert gas. Protect from moisture. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage. Harmful if inhaled. Harmful in contact with skin. Harmful if swallowed. May cause an allergic skin reaction. May cause respiratory irritation. May cause damage to organs through prolonged or repeated exposure. May cause long lasting harmful effects to aquatic life. Highly flammable liquid and vapor. May form explosive peroxides. Catches fire spontaneously if exposed to air. Suspected of causing cancer. May cause drowsiness or dizziness.
Formula Weight 99.97
Physical Form Solution
Quantity 800mL
Color Clear colorless to slightly amber
Packaging AcroSeal™ Glass Bottle
Density 0.8130g/mL
Linear Formula CH3OB(CH2CH3)2
Health Hazard 1 Danger
MDL Number MFCD00013240
Chemical Name or Material Diethylmethoxyborane
Molecular Formula C5H13BO
CAS 7397-46-8
Name Note 4M solution in THF
CAS 109-99-9

Specifications

Chemical Name or Material Diethylmethoxyborane
Name Note 4M solution in THF
CAS 7397-46-8
CAS 109-99-9
Molecular Formula C5H13BO
Linear Formula CH3OB(CH2CH3)2
Formula Weight 99.97
MDL Number MFCD00013240
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. Handle under inert gas. Protect from moisture. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
Causes severe skin burns and eye damage. Harmful if inhaled. Harmful in contact with skin. Harmful if swallowed. May cause an allergic skin reaction. May cause respiratory irritation. May cause damage to organs through prolonged or repeated exposure. May cause long lasting harmful effects to aquatic life. Highly flammable liquid and vapor. May form explosive peroxides. Catches fire spontaneously if exposed to air. Suspected of causing cancer. May cause drowsiness or dizziness.

SDS & Safety Documents