missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethylmethoxyborane, 4M solution in THF, AcroSeal™, ACROS Organics™
$114.13 - $114.13
Chemical Identifiers
CAS | 7397-46-8 |
---|---|
Molecular Formula | C5H13BO |
Molecular Weight (g/mol) | 99.97 |
MDL Number | MFCD00013240 |
InChI Key | FESAXEDIWWXCNG-UHFFFAOYSA-N |
Synonym | borinic acid, diethyl-, methyl ester, diethyl methoxy borane, diethyl methoxyborane, diethyl-methoxyborane, methoxy-diethylborane, methoxydiethyl-borane, methoxydiethylborane, methyl diethylborinate, unii-3ah40d7l25 |
PubChem CID | 522516 |
IUPAC Name | diethyl(methoxy)borane |
SMILES | B(CC)(CC)OC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC398381000
|
Thermo Fisher Scientific
AC398381000 |
100mL | AcroSeal™ Glass Bottle |
Each for $114.13
|
|
AC398388000
|
Thermo Fisher Scientific
AC398388000 |
800mL | AcroSeal™ Glass Bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Lewis acidChemical Identifiers
7397-46-8 | |
99.97 | |
FESAXEDIWWXCNG-UHFFFAOYSA-N | |
522516 | |
B(CC)(CC)OC |
C5H13BO | |
MFCD00013240 | |
borinic acid, diethyl-, methyl ester, diethyl methoxy borane, diethyl methoxyborane, diethyl-methoxyborane, methoxy-diethylborane, methoxydiethyl-borane, methoxydiethylborane, methyl diethylborinate, unii-3ah40d7l25 | |
diethyl(methoxy)borane |
Specifications
0.8130g/mL | |
AcroSeal™ Glass Bottle | |
Amber to Colorless | |
7397-46-8,109-99-9 | |
CH3OB(CH2CH3)2 | |
borinic acid, diethyl-, methyl ester, diethyl methoxy borane, diethyl methoxyborane, diethyl-methoxyborane, methoxy-diethylborane, methoxydiethyl-borane, methoxydiethylborane, methyl diethylborinate, unii-3ah40d7l25 | |
B(CC)(CC)OC | |
99.97 | |
99.97 | |
Diethylmethoxyborane, 4M solution in THF |
−17°C | |
0.813 | |
100mL | |
C5H13BO | |
MFCD00013240 | |
FESAXEDIWWXCNG-UHFFFAOYSA-N | |
diethyl(methoxy)borane | |
522516 | |
Solution |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause respiratory irritation.
May cause d
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger