missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl vinylphosphonate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 152150250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
for C-C Bond Formation (Olefination)Specifications
Diethyl vinylphosphonate | |
1.0600g/mL | |
>110°C | |
96% min. (GC) | |
C6H13O3P | |
1.4280 to 1.43 | |
25g | |
diethyl vinylphosphonate, vinylphosphonic acid diethyl ester, diethyl vinylphosphate, diethoxyphosphinylethene, phosphonic acid, vinyl-, diethyl ester, diethyl ethenylphosphonate, phosphonic acid, ethenyl-, diethyl ester, 1-ethenyl ethoxy phosphoryl oxyethane, acmc-1at5g | |
CCOP(=O)(OCC)C=C | |
164.14 | |
164.14 |
682-30-4 | |
202°C | |
Authentic | |
Glass bottle | |
(C2H5O)2P(O)CH=CH2 | |
MFCD00009079 | |
1.06 | |
DREPONDJUKIQLX-UHFFFAOYSA-N | |
1-[ethenyl(ethoxy)phosphoryl]oxyethane | |
69629 | |
97% |
Safety and Handling
EINECSNumber : 211-663-2
RUO â Research Use Only