missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyletrans-1,2-cyclopropanedicarboxylate, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC169180250

 View more versions of this product

Catalog No. AC169180250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Diethyl trans-1, 2-cyclopropanedicarboxylate
Authentic
1.4400 to 1.4420 (20°C, 589nm)
25g
97%
C3H4(CO2C2H5)2
diethyl trans-1,2-cyclopropanedicarboxylate, trans-diethyl cyclopropane-1,2-dicarboxylate, diethyl trans-cyclopropane-1,2-dicarboxylate, 1r,2r-diethyl cyclopropane-1,2-dicarboxylate, diethyl 1r,2r-cyclopropane-1,2-dicarboxylate, 1,2-cyclopropanedicarboxylic acid, diethyl ester, 1r,2r-rel, 1,2-cyclopropanedicarboxylicacid, 1,3-diethyl ester, 1r,2r-rel, 1s,2s-diethyl cyclopropane-1,2-dicarboxylate, diethyltrans-cyclopropane-1,2-dicarboxylate, trans-1,2-diethoxycarbonyl-cyclopropane
CCOC(=O)C1CC1C(=O)OCC
186.207
186.21
≥96.0%
1.0600g/mL
Glass bottle
Colorless to Yellow
3999-55-1
C9H14O4
MFCD00198002
SXLDHZFJMXLFJU-RNFRBKRXSA-N
diethyl (1R,2R)-cyclopropane-1,2-dicarboxylate
11715303
Liquid After Melting

Chemical Identifiers

3999-55-1
186.207
SXLDHZFJMXLFJU-RNFRBKRXSA-N
11715303
CCOC(=O)C1CC1C(=O)OCC
C9H14O4
MFCD00198002
diethyl trans-1,2-cyclopropanedicarboxylate, trans-diethyl cyclopropane-1,2-dicarboxylate, diethyl trans-cyclopropane-1,2-dicarboxylate, 1r,2r-diethyl cyclopropane-1,2-dicarboxylate, diethyl 1r,2r-cyclopropane-1,2-dicarboxylate, 1,2-cyclopropanedicarboxylic acid, diethyl ester, 1r,2r-rel, 1,2-cyclopropanedicarboxylicacid, 1,3-diethyl ester, 1r,2r-rel, 1s,2s-diethyl cyclopropane-1,2-dicarboxylate, diethyltrans-cyclopropane-1,2-dicarboxylate, trans-1,2-diethoxycarbonyl-cyclopropane
diethyl (1R,2R)-cyclopropane-1,2-dicarboxylate
SDS Documents