Learn More
Diethyl oxalate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 159180250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diethyl oxalate | |
0.01% max. (as oxalic acid) | |
1.0700g/mL | |
76°C | |
98.5% min. (GC) | |
C6H10O4 | |
C2H5O2CCO2C2H5 | |
25 g | |
01,250; 02,132; 07,106; 15,133 | |
15, 3142 | |
Solubility in water: insoluble. Other solubilities: soluble in methanol,ethanol,2-propanol and,acetone | |
CCOC(=O)C(=O)OCC | |
146.14 | |
146.14 | |
Liquid |
95-92-1 | |
-41.0°C | |
185.0°C | |
Authentic | |
Glass bottle | |
1.4085 to 1.4105 | |
MFCD00009119 | |
02, 535 | |
1.07 | |
ethyl oxalate, diethyl ethanedioate, ethanedioic acid, diethyl ester, oxalic acid, diethyl ester, oxalic ether, diethyloxalate, ethyl oxalate van, oxalic acid diethyl ester, unii-860m3zwf6j, diethylester kyseliny stavelove | |
WYACBZDAHNBPPB-UHFFFAOYSA-N | |
diethyl oxalate | |
7268 | |
99% |
Chemical Identifiers
95-92-1 | |
146.14 | |
WYACBZDAHNBPPB-UHFFFAOYSA-N | |
7268 | |
CCOC(=O)C(=O)OCC |
C6H10O4 | |
MFCD00009119 | |
ethyl oxalate, diethyl ethanedioate, ethanedioic acid, diethyl ester, oxalic acid, diethyl ester, oxalic ether, diethyloxalate, ethyl oxalate van, oxalic acid diethyl ester, unii-860m3zwf6j, diethylester kyseliny stavelove | |
diethyl oxalate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
Wear eye protection/face protection.
GHS Signal Word: Danger
EINECSNumber : 202-464-1
RUO – Research Use Only