Learn More
Diethylenetriaminepentaacetic acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1092630
Description
Diethylenetriaminepentaacetic acid is used as chelating agent and forms complexes with metal ions. It finds application as a magnetic resonance imaging (MRI) contrasting agent, since it improves the image by forming a complex with a gadolinium ion. It can also be used for treatment of radioactive materials such as plutonium, americium and other actinides. It is an active component of manganese and zinc fertilizers used in agrochemical industry. It acts as a color inhibitor and is used in the production of acrylon. It is also used in soaps as a water softener.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethylenetriaminepentaacetic acid is used as chelating agent and forms complexes with metal ions. It finds application as a magnetic resonance imaging (MRI) contrasting agent, since it improves the image by forming a complex with a gadolinium ion. It can also be used for treatment of radioactive materials such as plutonium, americium and other actinides. It is an active component of manganese and zinc fertilizers used in agrochemical industry. It acts as a color inhibitor and is used in the production of acrylon. It is also used in soaps as a water softener.
Solubility
Soluble in water and alkali.
Notes
Incompatible with strong oxidizing agents.
Specifications
Diethylenetriaminepentaacetic acid | |
∼220°C (decomposition) | |
Odorless | |
MFCD00004289 | |
1810219 | |
pentetic acid, diethylenetriaminepentaacetic acid, dtpa, detapac, complexon v, detarex, perma kleer, titriplex v, monaquest cai, hamp-ex acid | |
QPCDCPDFJACHGM-UHFFFAOYSA-N | |
2-[bis[2-[bis(carboxymethyl)amino]ethyl]amino]acetic acid | |
3053 | |
393.35 |
67-43-6 | |
1.56 | |
C14H23N3O10 | |
250 g | |
14,7125 | |
Soluble in water and alkali. | |
C(CN(CC(=O)O)CC(=O)O)N(CCN(CC(=O)O)CC(=O)O)CC(=O)O | |
393.349 | |
CHEBI:35739 | |
≥98% |
Chemical Identifiers
67-43-6 | |
393.349 | |
QPCDCPDFJACHGM-UHFFFAOYSA-N | |
3053 | |
2-[bis[2-[bis(carboxymethyl)amino]ethyl]amino]acetic acid |
C14H23N3O10 | |
MFCD00004289 | |
pentetic acid, diethylenetriaminepentaacetic acid, dtpa, detapac, complexon v, detarex, perma kleer, titriplex v, monaquest cai, hamp-ex acid | |
CHEBI:35739 | |
C(CN(CC(=O)O)CC(=O)O)N(CCN(CC(=O)O)CC(=O)O)CC(=O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 200-652-8
RTECSNumber : MB8205000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only