Please login to your online account to display your discounted pricing

Diethylenetriamine, 98+%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  114310050

 View more versions of this product

Catalog No. AC114310050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Formula Weight 103.17
Physical Form Liquid
Quantity 5mL
Color Clear
Density 0.9600g/mL
Linear Formula H2NCH2CH2NHCH2CH2NH2
Water 0.5% max. (K.F.)
Chemical Name or Material Diethylenetriamine
Assay Percent Range 98+%
Molecular Formula C4H13N3
CAS 111-40-0
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Fatal if inhaled.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Refractive Index 1.4826 to 1.4846 (20°C, 589nm)
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Diethylenetriamine
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C4H13N3
Linear Formula H2NCH2CH2NHCH2CH2NH2
Formula Weight 103.17
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Fatal if inhaled.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause respiratory irritation.

SDS & Safety Documents