missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethylene Glycol Monomethyl Ether (stabilized with BHT), TCI America™

Manufacturer:  TCI America M053725ML

 View more versions of this product

Catalog No. M053725ML



Specifications

Specifications

Diethylene Glycol Monomethyl Ether (stabilized with BHT)
Colorless
25mL
C5H12O3
2-2-methoxyethoxy ethanol, diethylene glycol monomethyl ether, methyl carbitol, methyl digol, methoxydiglycol, ethanol, 2-2-methoxyethoxy, methyl dioxitol, diethylene glycol methyl ether, dowanol dm, poly-solv dm
COCCOCCO
120.148
CHEBI:44836
Clear Liquid at 20°C
193°C
-70°C
111-77-3
MFCD00002871
SBASXUCJHJRPEV-UHFFFAOYSA-N
2-(2-methoxyethoxy)ethanol
8134
120.15
≥99.0% (GC)

Chemical Identifiers

111-77-3
120.148
SBASXUCJHJRPEV-UHFFFAOYSA-N
8134
2-(2-methoxyethoxy)ethanol
C5H12O3
MFCD00002871
2-2-methoxyethoxy ethanol, diethylene glycol monomethyl ether, methyl carbitol, methyl digol, methoxydiglycol, ethanol, 2-2-methoxyethoxy, methyl dioxitol, diethylene glycol methyl ether, dowanol dm, poly-solv dm
CHEBI:44836
COCCOCCO
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0422&(7)-0097

RTECSNumber : KL6125000

TSCA : Yes

Documents