missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethylene glycol monoethyl ether acetate, 99%

Manufacturer:  Alfa Aesar™ L13446AP

 View more versions of this product

Catalog No. AAL13446AP


 

Specifications

Description

Diethylene glycol monoethyl ether acetate; 99%; Alfa Aesar; 500mL; FW: 176.21; CH3CO2(CH2CH2O)2C2H5; MP: -25; BP: 218 to 219; CAS: 112-15-2

Applications
Diethylene glycol monoethyl ether acetate is used as a catalytic agent, petrochemical additive, coating auxiliary agent and in organic synthesis. It is mainly used as metal, furniture paint solvents and as organic intermediate.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with bases and oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
Description & Specifications

Specifications

Diethylene glycol monoethyl ether acetate
112-15-2
CH3CO2(CH2CH2O)2C2H5
MFCD00041928
1764643
95°C (203°F)
2-(2-Ethoxyethoxy)ethyl acetate
500mL
99%
176.21
218° to 219°C
−25°C
14,1800

Chemical Identifiers

112-15-2
176.21
1764643
2-(2-Ethoxyethoxy)ethyl acetate
CH3CO2(CH2CH2O)2C2H5
MFCD00041928
14,1800
Diethylene glycol monoethyl ether acetate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : KK8925000

TSCA : TSCA