Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethylene glycol monoethyl ether acetate, 99%


Quantity 100mL

Manufacturer: alfa aesar™  L13446AE

 View more versions of this product

Catalog No. AAL13446AE

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 176.21
Molecular Formula CH3CO2(CH2CH2O)2C2H5
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Assay 99%
Health Hazard 1 Warning
CAS 112-15-2
Beilstein 1764643
Merck Index 14,1800
MDL Number MFCD00041928
Synonym 2-(2-Ethoxyethoxy)ethyl acetate
Melting Point −25°C
RTECS Number KK8925000
Boiling Point 218° to 219°C
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Quantity 100mL
TSCA TSCA
Flash Point 95°C (203°F)
Chemical Name or Material Diethylene glycol monoethyl ether acetate

Specifications

Chemical Name or Material Diethylene glycol monoethyl ether acetate
CAS 112-15-2
Molecular Formula CH3CO2(CH2CH2O)2C2H5
Formula Weight 176.21
MDL Number MFCD00041928
Beilstein 1764643
Merck Index 14,1800
Synonym 2-(2-Ethoxyethoxy)ethyl acetate

Applications
Diethylene glycol monoethyl ether acetate is used as a catalytic agent, petrochemical additive, coating auxiliary agent and in organic synthesis. It is mainly used as metal, furniture paint solvents and as organic intermediate.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with bases and oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Containers which are opened must be carefully resealed and kept upright to prevent leakage.

Accessories

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.

SDS & Safety Documents