Please login to your online account to display your discounted pricing

Diethylene glycol monoethyl ether acetate, 97%, Alfa Aesar™

$24.42 - $153.28

Chemical Identifiers

Chemical Name or Material Diethylene glycol monoethyl ether acetate
CAS 112-15-2
Purity Grade Diethylene glycol monoethyl ether acetate, 97%
Physical Form Liquid
Assay Percent Range 97%
Molecular Formula CH3CO2(CH2CH2O)2C2H5
Formula Weight 176.21g/mol
MDL Number MFCD00041928
Synonym 2-(2-Ethoxyethoxy)ethyl acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA44170AD

 
alfa aesar
44170-AD
50mL Each for $24.42

AA44170K2

 
alfa aesar
44170-K2
1L Each for $153.28
Description & Specifications

Specifications

Quantity 50mL
Chemical Name or Material Diethylene glycol monoethyl ether acetate
CAS 112-15-2
Purity Grade Diethylene glycol monoethyl ether acetate, 97%
Physical Form Liquid
Assay Percent Range 97%
Molecular Formula CH3CO2(CH2CH2O)2C2H5
Formula Weight 176.21g/mol
MDL Number MFCD00041928
Synonym 2-(2-Ethoxyethoxy)ethyl acetate
Health Hazard 1 H319: Causes serious eye irritation.
EINECS Number 203-940-1
Safety and Handling

H319: Causes serious eye irritation.