missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethylene glycol divinyl ether, 98%, stab. 0.1% potassium hydroxide

$61.26 - $186.26

Chemical Identifiers

CAS 764-99-8
Molecular Formula C8H14O3
Formula Weight 158.2
MDL Number MFCD00054610
Beilstein 1752316
Synonym 1,5-Divinyloxy-3-oxapentane
Chemical Name or Material Diethylene glycol divinyl ether, stabilized 0.1% potassium hydroxide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1250118 SDS Alfa Aesar™
L1250118
50g Each for $186.26
AAL1250109 SDS Alfa Aesar™
L1250109
10g Each for $61.26
Specifications

Description

Applications
Diethylene glycol divinyl ether is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents and light.
Description & Specifications

Chemical Identifiers

764-99-8
158.2
1752316
Diethylene glycol divinyl ether, stabilized 0.1% potassium hydroxide
C8H14O3
MFCD00054610
1,5-Divinyloxy-3-oxapentane

Specifications

50g
C8H14O3
MFCD00054610
1752316
83°C (181°F)
Diethylene glycol divinyl ether, stabilized 0.1% potassium hydroxide
764-99-8
158.2
198° to 199°C
−21°C
1,5-Divinyloxy-3-oxapentane
98%

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : KN3850000

TSCA : TSCA