missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethylene glycol dibutyl ether, 99+%, pure, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 194982500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Diethylene glycol dibutyl ether | |
0.8800g/mL | |
118°C | |
99% min. (GC) | |
C12H26O3 | |
[CH3(CH2)3OCH2CH2]2O | |
0.88 | |
Solubility in water: moderate soluble (3g/L at 20°C). Other solubilities: soluble in chloroform | |
CCCCOCCOCCOCCCC | |
218.34 | |
2.39 mPa.s (20°C) | |
99+% |
112-73-2 | |
256.0°C | |
Authentic | |
Glass bottle | |
1.4225 to 1.4245 | |
250mL | |
diethylene glycol dibutyl ether, bis 2-butoxyethyl ether, dibutyl carbitol, butyl diglyme, 1-2-2-butoxyethoxy ethoxy butane, 5,8,11-trioxapentadecane, ether, bis butoxyethyl, 2,2'-dibutoxyethyl ether, ether, bis 2-butoxyethyl | |
KZVBBTZJMSWGTK-UHFFFAOYSA-N | |
1-[2-(2-butoxyethoxy)ethoxy]butane | |
8210 | |
218.34 | |
Pure |
Safety and Handling
EINECSNumber : 204-001-9
RUO â Research Use Only