Learn More
Diethylene glycol dibutyl ether, 99+%, Thermo Scientific Chemicals
Extraction of precious metals
Supplier: Thermo Scientific Chemicals 042150AE
Description
Extraction of precious metalsDiethylene glycol dibutyl ether is used as a solvent in Grignard reactions. It is also used as solvents in gold refining, decorative inks for ceramics and digital inks. It finds application in electrochemistry, gas absorption, extractant and high boiling reaction medium. It is also used in fuel, lubricant, textile and medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diethylene glycol dibutyl ether | |
-60°C | |
0.885 g/mL | |
117°C (243°F) | |
1.4235 | |
100 mL | |
Miscible with dimethyl sulfoxide,ethanol and acetone. Immiscible with water. | |
CCCCOCCOCCOCCCC | |
218.337 | |
218.74 | |
Liquid |
112-73-2 | |
Colorless | |
256°C | |
C12H26O3 | |
MFCD00009459 | |
diethylene glycol dibutyl ether, bis 2-butoxyethyl ether, dibutyl carbitol, butyl diglyme, 1-2-2-butoxyethoxy ethoxy butane, 5,8,11-trioxapentadecane, ether, bis butoxyethyl, 2,2'-dibutoxyethyl ether, ether, bis 2-butoxyethyl | |
KZVBBTZJMSWGTK-UHFFFAOYSA-N | |
1-[2-(2-butoxyethoxy)ethoxy]butane | |
8210 | |
≥99% |
Chemical Identifiers
112-73-2 | |
218.337 | |
KZVBBTZJMSWGTK-UHFFFAOYSA-N | |
8210 | |
CCCCOCCOCCOCCCC |
C12H26O3 | |
MFCD00009459 | |
diethylene glycol dibutyl ether, bis 2-butoxyethyl ether, dibutyl carbitol, butyl diglyme, 1-2-2-butoxyethoxy ethoxy butane, 5,8,11-trioxapentadecane, ether, bis butoxyethyl, 2,2'-dibutoxyethyl ether, ether, bis 2-butoxyethyl | |
1-[2-(2-butoxyethoxy)ethoxy]butane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 204-001-9
RTECSNumber : KN0350000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only