missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethylene glycol diacrylate, tech. 75%, stab., Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1068222

 View more versions of this product

Catalog No. AAL1068222



Description

Description

Diethylene glycol diacrylate is used as reactive diluent and cross linking agent of radiation curing system, and also may be used as crosslinking resins, plastics and rubber modifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol diacrylate is used as reactive diluent and cross linking agent of radiation curing system, and also may be used as crosslinking resins, plastics and rubber modifier.

Solubility
Slightly soluble in water (1-5g/100ml at 18°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to light. Incompatible with strong oxidizing agents.
Specifications

Specifications

Diethylene glycol diacrylate
4074-88-8
94°C (0.2 mmHg)
C10H14O5
MFCD00014939
UN2810
diethylene glycol diacrylate, oxydiethylene acrylate, oxydiethylene diacrylate, acrylic acid, oxydiethylene ester, tga 2, 2-propenoic acid, oxydi-2,1-ethanediyl ester, diacrylate diethylene glycol, unii-jk01t392ju, acrylic acid, 2-ethoxyethanol diester, di ethylene glycol diacrylate
LEJBBGNFPAFPKQ-UHFFFAOYSA-N
2-(2-prop-2-enoyloxyethoxy)ethyl prop-2-enoate
19996
75%
stabilized
1.118
>110°C (230°F)
1.462
100 g
1953711
Slightly soluble in water (1-5g/100ml at 18°C).
C=CC(=O)OCCOCCOC(=O)C=C
214.22
214.22
Technical

Chemical Identifiers

4074-88-8
214.22
LEJBBGNFPAFPKQ-UHFFFAOYSA-N
19996
C=CC(=O)OCCOCCOC(=O)C=C
C10H14O5
MFCD00014939
diethylene glycol diacrylate, oxydiethylene acrylate, oxydiethylene diacrylate, acrylic acid, oxydiethylene ester, tga 2, 2-propenoic acid, oxydi-2,1-ethanediyl ester, diacrylate diethylene glycol, unii-jk01t392ju, acrylic acid, 2-ethoxyethanol diester, di ethylene glycol diacrylate
2-(2-prop-2-enoyloxyethoxy)ethyl prop-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H311-H315-H319-H317
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P261-P264b-P272-P280-P302+P352-P305+P351+P338-P312-P333+P313-P361-P363-P501c

H311-H315-H317-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 223-791-6

RTECSNumber : AS9450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only