missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl isocyanomethylphosphonate, 97%, ACROS Organics™
Chemical Identifiers
CAS | 41003-94-5 |
---|---|
Molecular Formula | C6H12NO3P |
Molecular Weight (g/mol) | 177.14 |
MDL Number | MFCD00042951 |
InChI Key | ICURVXBGGDVHDT-UHFFFAOYSA-N |
Synonym | 1-ethoxy isocyanomethyl phosphoryl oxyethane, acmc-20al8q, diethoxycarbonyl methanisocyanide, diethoxyphosphinylmethyl isocyanide, diethyl isocyanomethyl phosphonate, diethyl isocyanomethylphosphonate, diethylisocyanomethylphosphonate, isocyanomethylphosphonic acid diethyl ester, isocyanomethylphosphononic acid diethyl ester |
PubChem CID | 2734699 |
IUPAC Name | 1-[ethoxy(isocyanomethyl)phosphoryl]oxyethane |
SMILES | CCOP(=O)(C[N+]#[C-])OCC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC369240010
|
Thermo Fisher Scientific
AC369240010 |
1mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC369240050
|
Thermo Fisher Scientific
AC369240050 |
5mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
for C-C Bond Formation (Olefination)Chemical Identifiers
41003-94-5 | |
177.14 | |
ICURVXBGGDVHDT-UHFFFAOYSA-N | |
2734699 | |
CCOP(=O)(C[N+]#[C-])OCC |
C6H12NO3P | |
MFCD00042951 | |
1-ethoxy isocyanomethyl phosphoryl oxyethane, acmc-20al8q, diethoxycarbonyl methanisocyanide, diethoxyphosphinylmethyl isocyanide, diethyl isocyanomethyl phosphonate, diethyl isocyanomethylphosphonate, diethylisocyanomethylphosphonate, isocyanomethylphosphonic acid diethyl ester, isocyanomethylphosphononic acid diethyl ester | |
1-[ethoxy(isocyanomethyl)phosphoryl]oxyethane |
Specifications
1.1050g/mL | |
Authentic | |
1.4310 to 1.4340 (20°C, 589nm) | |
84°C to 87°C (0.1mmHg) | |
1mL | |
96% min. (GC) | |
CNCH2P(O)(OC2H5)2 | |
14,135 | |
ICURVXBGGDVHDT-UHFFFAOYSA-N | |
1-[ethoxy(isocyanomethyl)phosphoryl]oxyethane | |
2734699 | |
Liquid | |
Diethyl isocyanomethylphosphonate |
>110°C | |
Glass bottle | |
1.105 | |
Colorless to Yellow | |
41003-94-5 | |
C6H12NO3P | |
MFCD00042951 | |
1-ethoxy isocyanomethyl phosphoryl oxyethane, acmc-20al8q, diethoxycarbonyl methanisocyanide, diethoxyphosphinylmethyl isocyanide, diethyl isocyanomethyl phosphonate, diethyl isocyanomethylphosphonate, diethylisocyanomethylphosphonate, isocyanomethylphosphonic acid diethyl ester, isocyanomethylphosphononic acid diethyl ester | |
CCOP(=O)(C[N+]#[C-])OCC | |
177.14 | |
177.14 | |
97% |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a
GHS Signal Word: Warning