missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyledimethylmalonate, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC162660500

 View more versions of this product

Catalog No. AC162660500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Diethyledimethylmalonate
Authentic
1.4114 to 1.4134 (20°C, 589nm)
50g
97%
C2H5O2CC(CH3)2CO2C2H5
02, II, 572
UELKSYXXNGHTSE-UHFFFAOYSA-N
diethyl 2,2-dimethylpropanedioate
15382
Liquid
0.99g/mL
Glass bottle
Colorless
1619-62-1
C9H16O4
MFCD00009128
diethyl 2,2-dimethylmalonate, diethyl dimethylmalonate, propanedioic acid, dimethyl-, diethyl ester, diethyldimethylmalonate, diethyl dimethyl malonate, 1,3-diethyl 2,2-dimethylpropanedioate, malonic acid, dimethyl-, diethyl ester, 2,2-dimethyl-malonic acid diethyl ester, zlchem 328, pubchem16321
CCOC(=O)C(C)(C)C(=O)OCC
188.223
188.22
≥96.0%

Chemical Identifiers

1619-62-1
188.223
UELKSYXXNGHTSE-UHFFFAOYSA-N
15382
CCOC(=O)C(C)(C)C(=O)OCC
C9H16O4
MFCD00009128
diethyl 2,2-dimethylmalonate, diethyl dimethylmalonate, propanedioic acid, dimethyl-, diethyl ester, diethyldimethylmalonate, diethyl dimethyl malonate, 1,3-diethyl 2,2-dimethylpropanedioate, malonic acid, dimethyl-, diethyl ester, 2,2-dimethyl-malonic acid diethyl ester, zlchem 328, pubchem16321
diethyl 2,2-dimethylpropanedioate
SDS Documents