missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl dimethylmalonate, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 1619-62-1 |
---|---|
Molecular Formula | C9H16O4 |
Molecular Weight (g/mol) | 188.22 |
MDL Number | MFCD00009128 |
InChI Key | UELKSYXXNGHTSE-UHFFFAOYSA-N |
Synonym | 1,3-diethyl 2,2-dimethylpropanedioate, 2,2-dimethyl-malonic acid diethyl ester, diethyl 2,2-dimethylmalonate, diethyl dimethyl malonate, diethyl dimethylmalonate, diethyldimethylmalonate, malonic acid, dimethyl-, diethyl ester, propanedioic acid, dimethyl-, diethyl ester, pubchem16321, zlchem 328 |
PubChem CID | 15382 |
IUPAC Name | diethyl 2,2-dimethylpropanedioate |
SMILES | CCOC(=O)C(C)(C)C(=O)OCC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC162660100
|
Thermo Fisher Scientific
162660100 |
10g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1619-62-1 | |
188.22 | |
UELKSYXXNGHTSE-UHFFFAOYSA-N | |
15382 | |
CCOC(=O)C(C)(C)C(=O)OCC |
C9H16O4 | |
MFCD00009128 | |
1,3-diethyl 2,2-dimethylpropanedioate, 2,2-dimethyl-malonic acid diethyl ester, diethyl 2,2-dimethylmalonate, diethyl dimethyl malonate, diethyl dimethylmalonate, diethyldimethylmalonate, malonic acid, dimethyl-, diethyl ester, propanedioic acid, dimethyl-, diethyl ester, pubchem16321, zlchem 328 | |
diethyl 2,2-dimethylpropanedioate |
Specifications
1619-62-1 | |
Colorless | |
71°C | |
Authentic | |
C2H5O2CC(CH3)2CO2C2H5 | |
02, II, 572 | |
1.4114 to 1.4134 | |
10g | |
0.99 | |
diethyl 2,2-dimethylpropanedioate | |
15382 | |
Liquid | |
Diethyl dimethylmalonate |
0.9900g/mL | |
192°C | |
97% | |
C9H16O4 | |
MFCD00009128 | |
Glass bottle | |
1,3-diethyl 2,2-dimethylpropanedioate, 2,2-dimethyl-malonic acid diethyl ester, diethyl 2,2-dimethylmalonate, diethyl dimethyl malonate, diethyl dimethylmalonate, diethyldimethylmalonate, malonic acid, dimethyl-, diethyl ester, propanedioic acid, dimethyl-, diethyl ester, pubchem16321, zlchem 328 | |
UELKSYXXNGHTSE-UHFFFAOYSA-N | |
CCOC(=O)C(C)(C)C(=O)OCC | |
188.22 | |
188.22 | |
97% |
Safety and Handling
EINECSNumber : 216-582-6
RUO – Research Use Only